

Cyclopentanecarbonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4524-93-0
:
Formula:C₆H₉ClO
MDL Number:MFCD00001370
:132.59 g/mol
Boiling Pt:54…56 °C (20 hPa)
Density:1.1002 g/cm³ (20 °C)
EINECS:224-856-1
UN:2920
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together