Order Entry
United Kingdom
ContactUsLinkComponent
Benzotriazole
Benzotriazole
Catalog # 705-2028
Supplier:  Hach
CAS Number:  
undefined
Benzotriazole
Catalog # 705-2028
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :119.13 g/mol
Merck Index:13,01109
Boiling Pt:350 °C (1013 hPa)
Melting Pt:99 °C
Density:1.34 g/cm³ (20 °C)
Flash Pt:170 °C (closed cup)
Storage Temperature:Ambient
EINECS:202-394-1
UN:2811
ADR:6.1,III
 :GHS02
 :GHS07
CAS Number:95-14-7
Formula:C₆H₅N₃
MDL Number:MFCD00005699