£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Benzotriazole
Benzotriazole ≥99%
Benzotriazole ≥99%
:
Thermo Fisher Scientific
Benzotriazole ≥99%
:
Thermo Fisher Scientific
:
95-14-7
:
:
C₆H₅N₃
:
MFCD00005699
:
119.13 g/mol
:
14,1108
:
204 °C (15 mmHg)
:
96…99 °C
:
1.360
:
212 °C (413 °F)
:
Ambient
:
202-394-1
:
2811
:
6.1,III
:
GHS07
msds_ALFAA15423
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
(3-Aminopropyl)triethoxysilane for synthesis, Sigma-Aldrich®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Potassium hydroxide solution 0.1 mol/l (0.1 N) in 2-propanol acc. to DIN 51558 part 1, AVS TITRINORM
: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
VWR® Cooling Bricks
: VWR Collection
5-Methylbenzotriazole, Sigma-Aldrich®
: Merck
Acrylonitrile ≥99% stabilised
: Thermo Fisher Scientific