Order Entry
United Kingdom
ContactUsLinkComponent
Coumarin
Irritant
Coumarin
Catalog # EHERC11735000
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Coumarin
Catalog # EHERC11735000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :146.15 g/mol
Merck Index:13,02588
Boiling Pt:298 °C (1013 hPa)
Melting Pt:67…69 °C
Density:0.935 g/cm³ (20 °C)
Flash Pt:162 °C
Storage Temperature:Ambient
EINECS:202-086-7
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:91-64-5
Formula:C₉H₆O₂
MDL Number:MFCD00006850