Order Entry
United Kingdom
ContactUsLinkComponent
2,2',3,3',4,4'-Hexachlorobiphenyl (PCB No. 128)
2,2',3,3',4,4'-Hexachlorobiphenyl (PCB No. 128)
Catalog # EHERC20012800
Supplier:  EHRENSTORFER
CAS Number:  
2,2',3,3',4,4'-Hexachlorobiphenyl (PCB No. 128)
Catalog # EHERC20012800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :360.88 g/mol
UN:3432
ADR:9,II
CAS Number:38380-07-3
Formula:C₁₂H₄Cl₆