
2-Mesitylenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:773-64-8
:
Formula:C₉H₁₁ClO₂S
MDL Number:MFCD00007434
:218.7 g/mol
Boiling Pt:150 °C (20 mmHg)
Melting Pt:53…56 °C
Storage Temperature:Ambient
EINECS:212-257-8
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ200
:GHS05
Frequently Bought Together