3,5-Dimethylbenzylamine
Catalog # ABCRMK120.0005
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:135.21 g/mol
EINECS:278-974-3
CAS Number:78710-55-1
Formula:C₉H₁₃N
MDL Number:MFCD00040753