Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethylbenzylamine
3,4-Dimethylbenzylamine
3,4-Dimethylbenzylamine
3,4-Dimethylbenzylamine
FLUO018617-5G
:
FLUOROCHEM
:
3,4-Dimethylbenzylamine
FLUO018617-5G
:
FLUOROCHEM
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
135.21 g/mol
:
203-034-6
:
102-48-7
:
C₉H₁₃N
:
MFCD00025577