3,4-Dimethylbenzylamine
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102-48-7
:
Formula:C₉H₁₃N
MDL Number:MFCD00025577
:135.21 g/mol
Storage Temperature:Ambient
EINECS:203-034-6
ADR:8,III
:GHS05
Related Information
Frequently Bought Together