3,5-Dimethyl-p-anisic acid
Catalog # FLUO022642-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.2 g/mol
Boiling Pt:313 °C (1013 hPa)
Melting Pt:196…198 °C
Density:1,253 g/cm³ (20 °C)
EINECS:244-441-9
CAS Number:21553-46-8
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00020309