
3,5-Dimethyl-p-anisic acid 98%
ACRO187130050
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Appearance:White to brown Crystalline solid
- Infrared spectrum:Conforms
- Melting point:190 to 194 °C
- Titration with NaOH:≥97.5 %
- UV:at 250 nm A: ≤0.3
- Relative density:0.805 to 0.812
- Residue after evaporation:≤25 ppm
- Acidity/alkalinity:Passes test
- Ethanol:95.1 to 96.9 % v/v (calculated)
- Volatile impurities:≤100 ppm (Methanol)
:
:180.2 g/mol
:313 °C (1013 hPa)
:190…194 °C
:1,253 g/cm³ (20 °C)
:244-441-9
:GHS07
:21553-46-8
:C₁₀H₁₂O₃
:MFCD00020309