Order Entry
United Kingdom
ContactUsLinkComponent
4,4'-Methylenedianiline
Health
Environment
4,4'-Methylenedianiline
Catalog # EHERC10648000
Supplier:  EHRENSTORFER
CAS Number:  
Health
Environment
4,4'-Methylenedianiline
Catalog # EHERC10648000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :198.27 g/mol
Merck Index:13,03000
Boiling Pt:398 °C (1013 hPa)
Melting Pt:91…92 °C
Density:1.15 g/cm³ (20 °C)
Flash Pt:220 °C
Storage Temperature:Ambient
EINECS:202-974-4
UN:2651
ADR:6.1,III
 :GHS08
 :GHS09
CAS Number:101-77-9
Formula:C₁₃H₁₄N₂
MDL Number:MFCD00007914