Triphenylmethane
Catalog # MOLE38673033-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:244.34 g/mol
Merck Index:12,09871
Boiling Pt:359 °C (1013 hPa)
Melting Pt:90…92 °C
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-275-0
CAS Number:519-73-3
Formula:C₁₉H₁₆
MDL Number:MFCD00004763