Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Benzil 99+%

Irritant
Benzil 99+%
Certificates
Benzil 99+%

3 Options Available Below

Product Details & Documents
Synonyms:  1,2-Diphenyl-1,2-ethanedione
  • CAS Number:134-81-6
  • Formula:(C₆H₅)C(O)C(O)(C₆H₅)
  • MDL Number:MFCD00003080
  • Molecular Weight:210.23 g/mol
  • Merck Index:13,01078
  • Boiling Pt:346…348 °C (760 mmHg)
  • Melting Pt:94…97 °C
  • Density:1,23 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-157-0

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    93 to 97 °C
  • GC:
    ≥99.0 %
  • Calcium (Ca):
    ≤100 ppm
  • Iron (Fe):
    ≤500 ppm
  • Potassium (K):
    ≤200 ppm
  • Sodium (Na):
    ≤500 ppm
  • Carbon compounds (as C):
    ≤0.01 %
  • Insoluble matter:
    ≤0.02 % (in dilute HCl)
  • Nitrogen compounds (as N):
    ≤0.002 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £19.50
    £19.50
    1 * 100 g
       £19.50

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £49.70
    £49.70
    1 * 500 g
       £49.70

  • Plastic bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - £158.00
    £158.00
    1 * 2,5 kg
       £158.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.