Order Entry
United Kingdom
ContactUsLinkComponent
Bromochlorophenol blue 95% (dye content), Sigma-Aldrich®
Bromochlorophenol blue 95% (dye content), Sigma-Aldrich®
  510998-5G
 :  
undefined
Bromochlorophenol blue 95% (dye content), Sigma-Aldrich®
  510998-5G
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

 

 

  • Appearance (Form):
    Powder
  • Infrared Spectrum:
    Conforms to Structure
  • Titration with AgNO3
    After Oxygen Combustion:
    94.5 - 105.5 %
  • Solubility (Turbidity)
    10 mg/ml, Ethanol:
    Clear
  • Solubility (Colour):
    Faint Orange to Very Dark Orange
  • Wavelength
    0.01 g/L, 0.1 N NaOH:
    586 - 590 nm
  • Extinction Coefficient:
    ≥70000
  • Wavelength:
    378 - 382 nm
  • Visual Interval
    pH 3.0 = Yellow:
    Pass
  • Visual Interval
    pH 4.6 = Purple:
    Pass

 

 

 : 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromo-6-chlorophenol] S,S-dioxide
 :
 :581.06 g/mol
 :603
 :250…251 °C
 :Room Temperature
 :219-861-0
 :2553-71-1
 :C₁₉H₁₀Br₂Cl₂O₅S
 :MFCD00047291
 :M-Clarity™ quality level = MQ100