p-Toluenesulphonyl chloride, Sigma-Aldrich®
: Merck
:98-59-9
:
:C₇H₇ClO₂S
:MFCD00007450
:190.65 g/mol
:13,09612
:265 °C (1013 hPa)
:67 °C
:1.49 g/cm³ (20 °C)
:128 °C (closed cup)
:Ambient
:202-684-8
:3261
:8,II
:GHS05
Frequently Bought Together