
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:100-51-6
:
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599
:108.14 g/mol
Merck Index:13,01126
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–15,3 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:101 °C
EINECS:202-859-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (on anhydrous substance):Min. 99.0 %
- Acidity:Max. 0.0008 meq/g
- Boiling point:204 - 208 °C
- Density (20/4):1.043 - 1.046
- Peroxide value:Max. 10
- Benzaldehyde:Max. 0.1 %
- Water:Max. 0.1 %
- Cl (Chloride):Max. 20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
20807_BENZYL ALCOHOL ANALYTICAL REAGENT_BSE_REV0.pdf
BSE/TSE Certificates
Frequently Bought Together