
Benzyl alcohol 98.0-100.5% NF, J.T.Baker®
Catalog # 9040-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:98.0 - 100.5 %
- NF - Assay:98.0 - 100.5 %
- NF - Clarity of Solution:Passes Test
- NF - Color of Solution:Passes Test
- NF - Identification:Passes Test
- NF - Peroxide Value:≤5
- NF - Refractive Index, n²ºD:1.538 - 1.541
- NF - Benzaldehyde and Other Related Substances - Benzaldehyde:≤0.15 %
- NF - Related Substances - Cyclohexylmethanol:≤0.10 %
- of Benzyl Alcohol:≤0.04 %
- NF - Benzaldehyde and Other Related Substances - Total Other Peaks≤0.04 %
- NF - Benzaldehyde and Other Related Substances - Total Other Peaks:≤0.3 %
- Greater Than RT of Benzyl Alcohol:≤0.3 %
- NF - Acidity:Passes Test
- NF - Residue on Evaporation:≤0.05 %
- NF - Benzene:≤2 ppm
Specification Test Results
About this item
:
:108.14 g/mol
Merck Index:13,01126
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–15.4…–15.4 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:100.40 °C
Storage Temperature:Ambient
EINECS:202-859-9
:GHS07
CAS Number:100-51-6
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599