Order Entry
United Kingdom
ContactUsLinkComponent
2-Methylanisole, Sigma-Aldrich®
2-Methylanisole, Sigma-Aldrich®
Catalog # SIALM29355-100ML
Supplier:  Merck
CAS Number:  
2-Methylanisole, Sigma-Aldrich®
Catalog # SIALM29355-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :122.17 g/mol
Boiling Pt:172 °C (1013 hPa)
Density:0.98 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,III
 :M-Clarity™ quality level = MQ100
EINECS:209-426-3
UN:3271
CAS Number:578-58-5
Formula:C₈H₁₀O
MDL Number:MFCD00008373