p-Toluidine, Supelco®
Supplier: Merck
Jump to:
About this item
Synonyms: 4-Aminotoluene, 4-Methylaniline, 4-Toluidine
- CAS Number:106-49-0
- Formula:C₇H₉N
- MDL Number:MFCD00007906
- Molecular Weight:107.16 g/mol
- Merck Index:12,09674
- Boiling Pt:201 °C (1013 hPa)
- Melting Pt:43 °C
- Density:1.046 g/cm³ (20 °C)
- Flash Pt:87 °C
- Storage Temperature:Ambient
- UN:3451
- ADR:6.1,II
- :M-Clarity™ quality level = MQ100
- EINECS:203-403-1
2 Options Available Below
Product Details & Documents
Synonyms: 4-Aminotoluene, 4-Methylaniline, 4-Toluidine
- CAS Number:106-49-0
- Formula:C₇H₉N
- MDL Number:MFCD00007906
- Molecular Weight:107.16 g/mol
- Merck Index:12,09674
- Boiling Pt:201 °C (1013 hPa)
- Melting Pt:43 °C
- Density:1.046 g/cm³ (20 °C)
- Flash Pt:87 °C
- Storage Temperature:Ambient
- UN:3451
- ADR:6.1,II
- :M-Clarity™ quality level = MQ100
- EINECS:203-403-1



