m-Toluidine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 108-44-1 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 85°(185°F) Refractive Index: 1.5670 Odour: Unpleasant Melting Point: -30° Boiling Point: 203-204° Beilstein Reference: 635944 Merck Reference: 14,9536 UN No.: 1708 MDL No.: MFCD00007808
Synonyms: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline
- CAS Number:108-44-1
- Formula:C₇H₉N
- MDL Number:MFCD00007808
- Molecular Weight:107.16 g/mol
- Merck Index:14,9536
- Boiling Pt:203…204 °C
- Melting Pt:–30 °C
- Density:0.992
- Flash Pt:85 °C (185 °F)
- Storage Temperature:Ambient
- EINECS:203-583-1
- UN:1708
- ADR:6.1,II
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 108-44-1 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 85°(185°F) Refractive Index: 1.5670 Odour: Unpleasant Melting Point: -30° Boiling Point: 203-204° Beilstein Reference: 635944 Merck Reference: 14,9536 UN No.: 1708 MDL No.: MFCD00007808
Synonyms: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline
- CAS Number:108-44-1
- Formula:C₇H₉N
- MDL Number:MFCD00007808
- Molecular Weight:107.16 g/mol
- Merck Index:14,9536
- Boiling Pt:203…204 °C
- Melting Pt:–30 °C
- Density:0.992
- Flash Pt:85 °C (185 °F)
- Storage Temperature:Ambient
- EINECS:203-583-1
- UN:1708
- ADR:6.1,II



