Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
m-Toluidine 99%

m-Toluidine 99%

Toxic
Health
Environment
m-Toluidine 99%
Certificates
m-Toluidine 99%

2 Options Available Below

Product Details & Documents
Synonyms:  3-Aminotoluene, 3-Toluidine, 3-Methylaniline
  • CAS Number:108-44-1
  • Formula:C₇H₉N
  • MDL Number:MFCD00007808
  • Molecular Weight:107.16 g/mol
  • Merck Index:13,09614
  • Boiling Pt:203…204 °C (760 mmHg)
  • Melting Pt:–30 °C
  • Density:0.98 g/cm³
  • Flash Pt:86 °C
  • Storage Temperature:Ambient
  • EINECS:203-583-1
  • UN:1708
  • ADR:6.1,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow to brown Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5655 to 1.5685 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Sulfate (SO4):
    ≤0.005 %
  • Iron (Fe):
    ≤0.002 %
  • Sodium (Na):
    ≤0.01 %
  • Heavy metals (as Pb):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - £22.10
    £22.10
    1 * 5 mL
       £22.10

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - £107.00
    £107.00
    1 * 1 L
       £107.00