3,4-Difluorobenzylamine 95%
Catalog # ACRO311530010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4920 to 1.4940 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
About this item
:
:143.14 g/mol
Boiling Pt:174 °C (1013 hPa)
Melting Pt:31 °C
Density:1.21 g/cm³
Flash Pt:79 °C
EINECS:276-503-6
UN:2735
ADR:8,III
:GHS05
CAS Number:72235-53-1
Formula:C₇H₇F₂N
MDL Number:MFCD00010145