
4-Methoxy-3-(trifluoromethyl)benzoyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98187-18-9
:
Formula:C₉H₆ClF₃O₂
MDL Number:MFCD01631576
:238.59 g/mol
Boiling Pt:244 °C (1013 hPa)
Melting Pt:75 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together