Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

3',4'-Dihydroxyacetophenone

Marketsource
Supplier:   FLUOROCHEM
3',4'-Dihydroxyacetophenone
Supplier:  FLUOROCHEM
Marketsource
Certificates
3',4'-Dihydroxyacetophenone

2 Options Available Below

Product Details & Documents
Synonyms:  1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol
  • CAS Number:1197-09-7
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00016439
  • Molecular Weight:152.15 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource