Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4'-Hydroxyacetophenone, Sigma-Aldrich®

4'-Hydroxyacetophenone, Sigma-Aldrich®

Supplier:   Merck
4'-Hydroxyacetophenone, Sigma-Aldrich®
Supplier:  Merck
Certificates
4'-Hydroxyacetophenone, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
  • CAS Number:99-93-4
  • Formula:C₈H₈O₂
  • MDL Number:MFCD00002359
  • Molecular Weight:136.15 g/mol
  • Boiling Pt:313 °C (1013 hPa)
  • Melting Pt:105…108 °C
  • Density:1.139 g/cm³ (25 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:202-802-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £68.90
    £68.90
    1 * 5 g
       £68.90

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £105.00
    £105.00
    1 * 100 g
       £105.00

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £205.00
    £205.00
    1 * 500 g
       £205.00