4'-Hydroxyacetophenone ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 99-93-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 166°(330°F) Melting Point: 106-108° Boiling Point: 286° Beilstein Reference: 774355 MDL No.: MFCD00002359
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:286 °C
- Melting Pt:106…108 °C
- Density:1.139 g/cm³ (25 °C)
- Flash Pt:166 °C (330 °F)
- Storage Temperature:Ambient
- EINECS:202-802-8
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 99-93-4 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 166°(330°F) Melting Point: 106-108° Boiling Point: 286° Beilstein Reference: 774355 MDL No.: MFCD00002359
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:286 °C
- Melting Pt:106…108 °C
- Density:1.139 g/cm³ (25 °C)
- Flash Pt:166 °C (330 °F)
- Storage Temperature:Ambient
- EINECS:202-802-8
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



