2'-Hydroxyacetophenone for synthesis, Sigma-Aldrich®
Catalog # 8.04310.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
Specification Test Results
- Solubility:0.2 g/l (20 °C)
- Melting Point:4 - 6 °C
- Molar Mass:136.15 g/mol
- Boiling Point:212 - 214 °C (1013 hPa)
- Vapor Pressure:0.27 hPa (20 °C)
- Flash Point:106 °C
- Refractive Index:1.5584 (20 °C, 589 nm)
- Explosion Limit:0.98 - 11.8 %(V)
- Density:1.13 g/cm3 (20 °C)
- Synonyms:2-Acetylphenol
- Hill Formula:C8H8O2
- Chemical Formula:2-(OH)C6H4COCH3
Specification Test Results
About this item
:
:136.15 g/mol
Boiling Pt:213 °C (1013 hPa)
Density:1.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-288-0
CAS Number:118-93-4
:MFCD00002219
Formula:C₈H₈O₂