4-Chloro-o-cresol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1570-64-5
:
Formula:C₇H₇ClO
MDL Number:MFCD00002321
:142.58 g/mol
Boiling Pt:220…225 °C
Melting Pt:45…48 °C
Density:1.20
Flash Pt:78 °C (172 °F)
Storage Temperature:Ambient
EINECS:216-381-3
UN:3437
ADR:6.1,II
:GHS06
:GHS05
:GHS09
Related Information
Frequently Bought Together