4-Chlorobenzyl alcohol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:873-76-7
:
Formula:C₇H₇ClO
MDL Number:MFCD00004652
:142.58 g/mol
Boiling Pt:234 °C (760 mmHg)
Melting Pt:69…72 °C
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:212-852-2
Specification Test Results
- Appearance:White to beige Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:69 to 72 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together