
4-Chloro-3-methylanisole ≥99%
Catalog # B23825.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:156.61 g/mol
Boiling Pt:26 °C (0.02 mmHg)
Density:1.14 g/cm³ (20 °C)
:GHS07
CAS Number:13334-71-9
Formula:C₈H₉ClO
MDL Number:MFCD00086700