Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4-methylanisole
2-Bromo-4-methylanisole, Sigma-Aldrich®
2-Bromo-4-methylanisole, Sigma-Aldrich®
SIAL480835-10G
:
Merck
:
2-Bromo-4-methylanisole, Sigma-Aldrich®
SIAL480835-10G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
201.06 g/mol
:
M-Clarity™ quality level = MQ100
:
244-705-3
:
GHS07
:
22002-45-5
:
C₈H₉BrO
:
MFCD00017783