α-Bromo-2,3,5,6-tetrafluorotoluene
Catalog # FLUO019466-10G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:243 g/mol
Boiling Pt:79…80 °C (1013 hPa)
EINECS:258-289-6
UN:3265
ADR:8,II
:GHS05
CAS Number:53001-73-3
Formula:C₇H₃BrF₄
MDL Number:MFCD00013549