ω-Bromo-3-methoxyacetophenone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Bromo-3'-methoxyacetophenone, Bromomethyl m-methoxyphenyl ketone, 3-Methoxyphenacyl bromide, 2-Bromo-1-(3-methoxyphenyl)ethanone, 2-Bromo-1-(3-methoxyphenyl)ethan-1-one, α-Bromo-3-methoxyacetophenone
CAS Number:5000-65-7
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00000199
:229.07 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:59…64 °C
Storage Temperature:Refrigerator
EINECS:225-666-1
UN:3261
ADR:8,II
:GHS07
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow-green Crystalline powder
- Infrared spectrum:Conforms
- Melting point:59 to 64 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together