Order Entry
United Kingdom
ContactUsLinkComponent
 
Phloroglucinol
Catalog # EDQMY0000493
Supplier:  EDQM
CAS Number:  
Phloroglucinol
Catalog # EDQMY0000493
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    210 mg
  • Seq:
    2545

Specifications

About this item

 :
 :126.11 g/mol
Merck Index:12,07482
Boiling Pt:400 °C (1013 hPa)
Melting Pt:217…220 °C
Density:1.46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-611-2
 :GHS08
CAS Number:108-73-6
Formula:C₆H₆O₃
MDL Number:MFCD00002286