Order Entry
United Kingdom
ContactUsLinkComponent
Sulphanilic acid
Sulphanilic acid
Catalog # ROTH4496.1
Supplier:  Roth Carl
CAS Number:  
undefined
Sulphanilic acid
Catalog # ROTH4496.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :173.19 g/mol
Merck Index:13,09012
Boiling Pt:363 °C (1013 hPa)
Melting Pt:279 °C
Density:1.485 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:204-482-5
 :GHS07
CAS Number:121-57-3
Formula:4-(H₂N)C₆H₄SO₃H
MDL Number:MFCD00007886