Order Entry
United Kingdom
ContactUsLinkComponent
Sulphanilic acid ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Irritant
Sulphanilic acid ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates
undefined
Irritant
Sulphanilic acid ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    10 g/l (20 °C)
  • Melting Point:
    288 °C decomposes
  • Molar Mass:
    173.18 g/mol
  • Bulk Density:
    620 kg/m3
  • Density:
    1.485 g/cm3 (25 °C)
  • pH:
    2.5 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    400 °C

Specification Test Results

Related Information

Frequently Bought Together