Order Entry
United Kingdom
ContactUsLinkComponent
Sulphanilic acid, Supelco®
Irritant
Sulphanilic acid, Supelco®
Catalog # 1.00684.0005
Supplier:  Merck
CAS Number:  
undefined
Irritant
Sulphanilic acid, Supelco®
Catalog # 1.00684.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Solubility:
    10 g/l (20 °C)
  • Melting Point:
    288 °C decomposes
  • Molar Mass:
    173.18 g/mol
  • Bulk Density:
    620 kg/m3
  • Density:
    1.485 g/cm3 (25 °C)
  • pH:
    2.5 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    400 °C

Specification Test Results

About this item

 :
 :173.19 g/mol
Merck Index:13,09012
Boiling Pt:363 °C (1013 hPa)
Melting Pt:279 °C
Density:1.485 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:204-482-5
 :GHS07
CAS Number:121-57-3
Formula:4-(H₂N)C₆H₄SO₃H
MDL Number:MFCD00007886