Order Entry
United Kingdom
ContactUsLinkComponent
2-Fluorobenzenesulphonyl chloride
2-Fluorobenzenesulphonyl chloride
Catalog # MAYBKM01957DA
Supplier:  Thermo Scientific
CAS Number:  
2-Fluorobenzenesulphonyl chloride
Catalog # MAYBKM01957DA
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.61 g/mol
Boiling Pt:242 °C (1013 hPa)
Density:1.47 g/cm³ (20 °C)
UN:3261
ADR:8,II
CAS Number:2905-21-7
Formula:C₆H₄ClFO₂S
MDL Number:MFCD00042285