
3,4,5-Trifluorobenzoyl chloride ≥97%
Catalog # L19188.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:17787-26-7
:177787-26-7
Formula:C₇H₂ClF₃O
MDL Number:MFCD00083526
:194.54 g/mol
Boiling Pt:186 °C (1013 hPa)
Melting Pt:41 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05