4-(Trifluoromethoxy)phenylboronic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:139301-27-2
:
Formula:C₇H₆BF₃O₃
MDL Number:MFCD01074648
:205.93 g/mol
Melting Pt:123…127 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together