O-Acetylsalicylic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-78-2
:
Formula:2-(CH₃CO₂)C₆H₄CO₂H
MDL Number:MFCD00002430
:180.16 g/mol
Merck Index:13,00856
Boiling Pt:140 °C (1013 hPa)
Melting Pt:138…140 °C
Density:1.39 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:250 °C
EINECS:200-064-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together