1-Chloro-4-nitrobenzene, Sigma-Aldrich®
Catalog # SIALC59122-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Merck Index:13,02170
Boiling Pt:239 °C (1013 hPa)
Melting Pt:83 °C
Density:1.52 g/cm³ (20 °C)
Flash Pt:127 °C
ADR:6.1,II
:M-Clarity™ quality level = MQ100
:157.56 g/mol
EINECS:202-809-6
UN:1578
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:100-00-5
Formula:C₆H₄ClNO₂
MDL Number:MFCD00007285