2-Amino-4-chlorophenol
Catalog # EHERC10199500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
- Environmentally Preferable:
Specifications
About this item
Synonyms: 5-Chloro-2-hydroxyaniline
:
:143.57 g/mol
EINECS:202-458-9
UN:2673
:GHS07
Storage Temperature:Ambient
CAS Number:95-85-2
Formula:C₆H₆ClNO
MDL Number:MFCD00007694
Melting Pt:140…143 °C
ADR:6.1 II