2-Chloro-4-fluorobenzenesulphonyl chloride
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:85958-57-2
:
Formula:C₆H₃Cl₂FO₂S
MDL Number:MFCD00052957
:229.06 g/mol
Boiling Pt:88…90 °C (1013 hPa)
UN:3265
ADR:8,II
Frequently Bought Together