
2-Chloro-4-fluorobenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:85958-57-2
:
Formula:C₆H₃Cl₂FO₂S
MDL Number:MFCD00052957
:229.06 g/mol
Boiling Pt:243…244 °C
Melting Pt:28…31 °C
Density:1.57
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together