Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-2,6-difluorobenzoyl chloride
3-Chloro-2,6-difluorobenzoyl chloride ≥97%
3-Chloro-2,6-difluorobenzoyl chloride ≥97%
L18244.03
:
Thermo Fisher Scientific
:
3-Chloro-2,6-difluorobenzoyl chloride ≥97%
L18244.03
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
261762-43-0
:
:
C₇H₂Cl₂F₂O
:
MFCD01631324
:
210.99 g/mol
:
222 °C (1013 hPa)
:
70 °C
:
Ambient
:
3265
:
8,II
:
GHS05