
4-Chlorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:74-11-3
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00002531
:156.57 g/mol
Merck Index:13,02144
Boiling Pt:287 °C (1013 hPa)
Melting Pt:123 °C
Density:1.535 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:238 °C
EINECS:200-805-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together