Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2,4-Dichlorophenylboronic acid 97%

2,4-Dichlorophenylboronic acid 97%

Irritant
2,4-Dichlorophenylboronic acid 97%
Certificates
2,4-Dichlorophenylboronic acid 97%

2 Options Available Below

Product Details & Documents
Synonyms:  Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid
  • CAS Number:68716-47-2
  • Formula:C₆H₅BCl₂O₂
  • MDL Number:MFCD00013930
  • Molecular Weight:190.82 g/mol
  • Melting Pt:240…245 °C

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • Titration with NaOH:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £17.50
    £17.50
    1 * 1 g
       £17.50

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £58.50
    £58.50
    1 * 5 g
       £58.50

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.