
5-Nitrosalicylaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:97-51-8
:
Formula:C₇H₅NO₄
MDL Number:MFCD00007337
:167.12 g/mol
Melting Pt:123…126 °C
:M-Clarity™ quality level = MQ100
EINECS:202-587-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together